N-(4-Fluorbenzoyl)morpholin
Lieferant: COMBI-BLOCKS
CA-4532-5G
CA-4532-25G
CA-4532-1G
COBBCA-4532-5GEA
223
EUR
COBBCA-4532-5G
COBBCA-4532-25G
COBBCA-4532-1G
N-(4-Fluorbenzoyl)morpholin
N-(4-Fluorbenzoyl)morpholin
Formel:
C₁₁H₁₂FNO₂ Molecular Weight: 209.22 g/mol |
MDL:
MFCD00451910 CAS-Nummer: 1978-65-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...