N-(3-Brom-4-fluorbenzoyl)morpholin
Lieferant: COMBI-BLOCKS
Synonyme:
(3-Bromo-4-fluorophenyl)(morpholino)methanone
CA-4610-5G
CA-4610-25G
CA-4610-1G
COBBCA-4610-5GEA
186
EUR
COBBCA-4610-5G
COBBCA-4610-25G
COBBCA-4610-1G
N-(3-Brom-4-fluorbenzoyl)morpholin
N-(3-Brom-4-fluorbenzoyl)morpholin
Formel:
C₁₁H₁₁BrFNO₂ Molecular Weight: 288,12 g/mol |
MDL:
MFCD11053798 CAS-Nummer: 1007207-89-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...