N-(4-Brom-2-fluorbenzoyl)morpholin

Lieferant: COMBI-BLOCKS

Synonyme: 4-(4-Bromo-2-fluorobenzoyl)morpholine

CA-4760-5G CA-4760-1G
COBBCA-4760-5GEA 884 EUR
COBBCA-4760-5G COBBCA-4760-1G
N-(4-Brom-2-fluorbenzoyl)morpholin
N-(4-Brom-2-fluorbenzoyl)morpholin
Formel: C₁₁H₁₁BrFNO₂
Molecular Weight: 288.12 g/mol
MDL: MFCD09945068
CAS-Nummer: 924642-61-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR