4-Brom-2-thiophencarbaldehyd

Lieferant: COMBI-BLOCKS

Synonyme: 4-Bromo-2-thiophenecarboxaldehyde, 4-Bromothiophene-2-carboxaldehyde

CS-9092-5G CS-9092-25G CS-9092-100G
COBBCS-9092-5GEA 0 EUR
COBBCS-9092-5G COBBCS-9092-25G COBBCS-9092-100G
4-Brom-2-thiophencarbaldehyd
4-Brom-2-thiophencarbaldehyd
Formel: C₅H₃BrOS
Molecular Weight: 191.05 g/mol
MDL: MFCD00005431
CAS-Nummer: 18791-75-8
EINECS: 242-577-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR