4-(2-Brom-4-nitrophenyl)morpholin

Lieferant: COMBI-BLOCKS

HC-2686-5G HC-2686-1G
COBBHC-2686-5GEA 0 EUR
COBBHC-2686-5G COBBHC-2686-1G
4-(2-Brom-4-nitrophenyl)morpholin
4-(2-Brom-4-nitrophenyl)morpholin
Formel: C₁₀H₁₁BrN₂O₃
MDL: MFCD02081938
CAS-Nummer: 477846-96-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR