3-Methyl-2-thenoesäure

Lieferant: COMBI-BLOCKS

Synonyme: 3-Methylthiophene-2-carboxylic acid

HI-1313-25G HI-1313-100G HI-1313-5G
COBBHI-1313-25GEA 116 EUR
COBBHI-1313-25G COBBHI-1313-100G COBBHI-1313-5G
3-Methyl-2-thenoesäure
3-Methyl-2-thenoesäure
Formel: C₆H₆O₂S
Molecular Weight: 142,18 g/mol
MDL: MFCD00005438
CAS-Nummer: 23806-24-8
EINECS: 245-894-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR