4-Brom-3-thiophencarbaldehyd

Lieferant: COMBI-BLOCKS

HI-1644-1G HI-1644-5G
COBBHI-1644-1GEA 122 EUR
COBBHI-1644-1G COBBHI-1644-5G
4-Brom-3-thiophencarbaldehyd
4-Brom-3-thiophencarbaldehyd
Formel: C₅H₃BrOS
Molecular Weight: 191.05 g/mol
MDL: MFCD01318960
CAS-Nummer: 18791-78-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR