3-Methoxyphenethylamin

Lieferant: COMBI-BLOCKS

Synonyme: 2-(3-Methoxyphenyl)ethylamine

OS-7450-25G OS-7450-5G
COBBOS-7450-25GEA 127 EUR
COBBOS-7450-25G COBBOS-7450-5G
3-Methoxyphenethylamin
3-Methoxyphenethylamin
Formel: C₉H₁₃NO
Molecular Weight: 151.21 g/mol
MDL: MFCD00008187
CAS-Nummer: 2039-67-0
EINECS: 218-017-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR