Metanilgelb
Lieferant: EHRENSTORFER
Synonyme:
Acid Yellow 36, 3-(4-Anilinophenylazo)benzenesulfonic acid sodium salt
C10029500
EHERC10029500EA
158
EUR
EHERC10029500
Metanilgelb
Metanilgelb
Formel:
C₁₈H₁₄N₃NaO₃S Molecular Weight: 375.38 g/mol |
MDL:
MFCD00007486 CAS-Nummer: 587-98-4 EINECS: 209-608-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...