(±)-4-Brom-α-methylbenzylamin
Lieferant: FLUOROCHEM
Synonyme:
4-Bromo-α-methylbenzylamine, (+/-)-1-(4-Bromophenyl)ethylamine, (±)-4-Bromo-α-methylbenzylamine
001101-5G
001101-25G
FLUO001101-5GEA
45.9
EUR
FLUO001101-5G
FLUO001101-25G
(±)-4-Brom-α-methylbenzylamin
(±)-4-Brom-α-methylbenzylamin
Formel:
C₈H₁₀BrN Molecular Weight: 200,08 g/mol |
MDL:
MFCD00025548 CAS-Nummer: 24358-62-1 EINECS: 246-200-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...