(±)-4-Fluor-α-methylbenzylamin

Lieferant: FLUOROCHEM

Synonyme: DL-4-Fluoro-α-methylbenzylamine, 1-(4-Fluorophenyl)ethylamine

001185-25G 001185-5G
FLUO001185-25GEA 303 EUR
FLUO001185-25G FLUO001185-5G
(±)-4-Fluor-α-methylbenzylamin
(±)-4-Fluor-α-methylbenzylamin
Formel: C₈H₁₀FN
Molecular Weight: 139.17 g/mol
MDL: MFCD00041323
CAS-Nummer: 403-40-7
EINECS: 206-958-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR