4-Brom-2-(trifluormethoxy)thiophenol
Lieferant: FLUOROCHEM
004302-1G
004302-5G
FLUO004302-1GEA
0
EUR
FLUO004302-1G
FLUO004302-5G
4-Brom-2-(trifluormethoxy)thiophenol
4-Brom-2-(trifluormethoxy)thiophenol
Formel:
C₇H₄BrF₃OS |
MDL:
MFCD00203474 CAS-Nummer: 175278-15-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...