2-Brom-3,3,3-trifluorpropen
Lieferant: FLUOROCHEM
Synonyme:
2-Bromo-3,3,3-trifluoro-1-propene
004309-5G
004309-25G
FLUO004309-5GEA
121
EUR
FLUO004309-5G
FLUO004309-25G
2-Brom-3,3,3-trifluorpropen
2-Bromo-3,3,3-trifluoropropene
Formel:
C₃H₂BrF₃ |
MDL:
MFCD00077469 CAS-Nummer: 1514-82-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...