3',5'-Difluor-4'-hydroxypropiophenon
Lieferant: FLUOROCHEM
004817-1G
004817-5G
FLUO004817-1GEA
87.9
EUR
FLUO004817-1G
FLUO004817-5G
3',5'-Difluor-4'-hydroxypropiophenon
3',5'-Difluor-4'-hydroxypropiophenon
Formel:
C₉H₈F₂O₂ Molecular Weight: 186.16 g/mol |
MDL:
MFCD00061287 CAS-Nummer: 178374-78-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...