3,5-Bis(trifluormethyl)benzamidoxim

Lieferant: FLUOROCHEM

Synonyme: N'-Hydroxy-3,5-bis(trifluoromethyl)benzimidamide

CFF257.10 006002-5G 006002-1G
CHMPCFF257.10EA 321 EUR
CHMPCFF257.10 FLUO006002-5G FLUO006002-1G
3,5-Bis(trifluormethyl)benzamidoxim
3,5-Bis(trifluormethyl)benzamidoxim
Formel: C₉H₆F₆N₂O
MDL: MFCD00068188
CAS-Nummer: 72111-09-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR