3',5'-Bis(trifluormethyl)propiophenon

Lieferant: FLUOROCHEM

Synonyme: 1-(3,5-Bis(trifluoromethyl)phenyl)propan-1-one

006038-1G 006038-5G 006038-25G
FLUO006038-1GEA 31.1 EUR
FLUO006038-1G FLUO006038-5G FLUO006038-25G
3',5'-Bis(trifluormethyl)propiophenon
3',5'-Bis(trifluormethyl)propiophenon
Formel: C₁₁H₈F₆O
Molecular Weight: 270.17 g/mol
MDL: MFCD00009911
CAS-Nummer: 85068-34-4
EINECS: 285-296-1
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR