trans-Perfluordecahydronaphthalin

Lieferant: FLUOROCHEM

Synonyme: trans-Perfluorodecalin

007016-5G 007016-1G
FLUO007016-5GEA 0 EUR
FLUO007016-5G FLUO007016-1G
trans-Perfluordecahydronaphthalin
trans-Perfluordecahydronaphthalin
Formel: C₁₀F₁₈
Molecular Weight: 462.08
MDL: MFCD00153232
CAS-Nummer: 60433-12-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR