4-Chlorphenethylamin

Lieferant: FLUOROCHEM

Synonyme: 2-(4-Chlorophenyl)ethylamine

008894-5G 008894-25G
FLUO008894-5GEA 0 EUR
FLUO008894-5G FLUO008894-25G
4-Chlorphenethylamin
4-Chlorphenethylamin
Formel: C₈H₁₀ClN
Molecular Weight: 155.63 g/mol
MDL: MFCD00008191
CAS-Nummer: 156-41-2
EINECS: 205-853-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR