3-Fluorphenethylbromid

Lieferant: FLUOROCHEM

Synonyme: 1-(2-Bromoethyl)-3-fluorobenzene

009225-1G 009225-5G
FLUO009225-1GEA 55.4 EUR
FLUO009225-1G FLUO009225-5G
3-Fluorphenethylbromid
3-Fluorphenethylbromid
Formel: C₈H₈BrF
Molecular Weight: 203,05 g/mol
MDL: MFCD03095397
CAS-Nummer: 25017-13-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR