2-Iod-3,3,3-trifluorpropen

Lieferant: FLUOROCHEM

010325-1G 010325-5G
FLUO010325-1GEA 0 EUR
FLUO010325-1G FLUO010325-5G
2-Iod-3,3,3-trifluorpropen
2-Iodo-3,3,3-trifluoropropene
Formel: C₃H₂F₃I
Molecular Weight: 221.95 g/mol
MDL: MFCD02183571
CAS-Nummer: 107070-09-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR