2-Benzo[b]thiophenboronsäure

Lieferant: CHEMPUR

Synonyme: Benzo[b]thiophen-2-ylboronic acid, 2-Benzothienylboronic acid

FL10979.POR 010979-5G
CHMPFL10979.POREA 0 EUR
CHMPFL10979.POR FLUO010979-5G
2-Benzo[b]thiophenboronsäure
2-Benzo[b]thiophenboronsäure
Formel: C₈H₇BO₂S
Molecular Weight: 178,02 g/mol
MDL: MFCD01075674
CAS-Nummer: 98437-23-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR