4-(Methylthio)phenylboronsäure

Lieferant: FLUOROCHEM

Synonyme: 4-(Methylthio)benzeneboronic acid, 4-Thioanisoleboronic acid

011008-1G 011008-5G
FLUO011008-1GEA 31.1 EUR
FLUO011008-1G FLUO011008-5G
4-(Methylthio)phenylboronsäure
4-(Methylthio)phenylboronsäure
Formel: C₇H₉BO₂S
Molecular Weight: 168.02 g/mol
MDL: MFCD00093410
CAS-Nummer: 98546-51-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR