4-Methyl-2-thiophenboronsäure
Lieferant: FLUOROCHEM
Synonyme:
(4-Methylthiophen-2-yl)boronic acid, 4-Methylthiophene-2-boronic acid
011050-5G
011050-1G
FLUO011050-5GEA
209
EUR
FLUO011050-5G
FLUO011050-1G
4-Methyl-2-thiophenboronsäure
4-Methyl-2-thiopheneboronic acid
Formel:
C₅H₇BO₂S Molecular Weight: 141.99 g/mol |
MDL:
MFCD01319040 CAS-Nummer: 162607-15-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...