(3,4-Dimethoxyphenyl)essigsäurehydrazid

Lieferant: FLUOROCHEM

011157-50G 011157-10G
FLUO011157-50GEA 381 EUR
FLUO011157-50G FLUO011157-10G
(3,4-Dimethoxyphenyl)essigsäurehydrazid
(3,4-Dimethoxyphenyl)essigsäurehydrazid
Formel: C₁₀H₁₄N₂O₃
Molecular Weight: 210.23 g/mol
MDL: MFCD00051701
CAS-Nummer: 60075-23-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR