(±)-2-Fluor-α-methylbenzylamin

Lieferant: CHEMPUR

Synonyme: 1-(2-Fluorophenyl)ethylamine

FL13377.POR 013377-1G
CHMPFL13377.POREA 0 EUR
CHMPFL13377.POR FLUO013377-1G
(±)-2-Fluor-α-methylbenzylamin
(±)-2-Fluor-α-methylbenzylamin
Formel: C₈H₁₀FN
Molecular Weight: 139,17 g/mol
MDL: MFCD04038307
CAS-Nummer: 74788-44-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR