4-(Trifluormethyl)benzamidoxim

Lieferant: FLUOROCHEM

CFF239.10 017291-1G 017291-25G 017291-5G
CHMPCFF239.10EA 321 EUR
CHMPCFF239.10 FLUO017291-1G FLUO017291-25G FLUO017291-5G
4-(Trifluormethyl)benzamidoxim
4-(Trifluormethyl)benzamidoxim
Formel: C₈H₇F₃N₂O
Molecular Weight: 204.15 g/mol
MDL: MFCD00053042
CAS-Nummer: 22179-86-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR