3-(Trifluormethyl)benzamidoxim

Lieferant: FLUOROCHEM

Synonyme: N'-Hydroxy-3-(trifluoromethyl)-benzenecarboximidamide, N-Hydroxy-3-(trifluoromethyl)benzimidamide

CFF256.10 CFF256.50 017306-10G 017306-25G
CHMPCFF256.10EA 321 EUR
CHMPCFF256.10 CHMPCFF256.50 FLUO017306-10G FLUO017306-25G
3-(Trifluormethyl)benzamidoxim
3-(Trifluormethyl)benzamidoxim
Formel: C₈H₇F₃N₂O
Molecular Weight: 204,15 g/mol
MDL: MFCD02647312
CAS-Nummer: 40067-80-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR