3-Methylbenzamidoxim
Lieferant: FLUOROCHEM
Synonyme:
N-Hydroxy-3-methylbenzimidamide
CFF268.5
CFF268.25
017315-5G
017315-10G
CHMPCFF268.5EA
532
EUR
CHMPCFF268.5
CHMPCFF268.25
FLUO017315-5G
FLUO017315-10G
3-Methylbenzamidoxim
3-Methylbenzamidoxim
Formel:
C₈H₁₀N₂O |
MDL:
MFCD00833266 CAS-Nummer: 40067-82-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...