4-Methylbenzamidoxim

Lieferant: FLUOROCHEM

Synonyme: N-Hydroxy-4-methylbenzimidamide

CFF269.5 CFF269.25 017316-5G 017316-10G
CHMPCFF269.5EA 532 EUR
CHMPCFF269.5 CHMPCFF269.25 FLUO017316-5G FLUO017316-10G
4-Methylbenzamidoxim
4-Methylbenzamidoxim
Formel: C₈H₁₀N₂O
MDL: MFCD00019952
CAS-Nummer: 19227-13-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR