Methyl-3-chlor-2-thiophencarboxylat

Lieferant: FLUOROCHEM

Synonyme: Methyl 3-chlorothiophene-2-carboxylate

CFF380.5 CFF380.10 CFF380.25 017406-5G 017406-10G 017406-25G
CHMPCFF380.5EA 348 EUR
CHMPCFF380.5 CHMPCFF380.10 CHMPCFF380.25 FLUO017406-5G FLUO017406-10G FLUO017406-25G
Methyl-3-chlor-2-thiophencarboxylat
Methyl-3-chlor-2-thiophencarboxylat
Formel: C₆H₅ClO₂S
Molecular Weight: 176.62 g/mol
MDL: MFCD00068135
CAS-Nummer: 88105-17-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR