5-Methylbenzofurazan

Lieferant: FLUOROCHEM

CFF513.5 CFF513.25 017519-5G 017519-25G 017519-1G
CHMPCFF513.5EA 348 EUR
CHMPCFF513.5 CHMPCFF513.25 FLUO017519-5G FLUO017519-25G FLUO017519-1G
5-Methylbenzofurazan
5-Methylbenzofurazan
Formel: C₇H₆N₂O
Molecular Weight: 134.14 g/mol
MDL: MFCD00068073
CAS-Nummer: 20304-86-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR