4-Nitrophenethylbromid

Lieferant: FLUOROCHEM

Synonyme: 2-(4-Nitrophenyl)-1-bromoethane, 1-(2-Bromoethyl)-4-nitrobenzene

019260-5G 019260-25G
FLUO019260-5GEA 0 EUR
FLUO019260-5G FLUO019260-25G
4-Nitrophenethylbromid
4-Nitrophenethylbromid
Formel: C₈H₈BrNO₂
Molecular Weight: 230.06 g/mol
MDL: MFCD00007386
CAS-Nummer: 5339-26-4
EINECS: 226-271-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR