4-Brombenzamidoxim

Lieferant: FLUOROCHEM

Synonyme: 4-Bromo-N-hydroxybenzimidamide

021899-5G 021899-25G
FLUO021899-5GEA 122 EUR
FLUO021899-5G FLUO021899-25G
4-Brombenzamidoxim
4-Brombenzamidoxim
Formel: C₇H₇BrN₂O
Molecular Weight: 215.05 g/mol
MDL: MFCD00019951
CAS-Nummer: 19227-14-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR