Boc-DL-Pro-OH

Lieferant: FLUOROCHEM

Synonyme: 1-(tert-Butoxycarbonyl)pyrrolidine-2-carboxylic acid, N-Boc-DL-proline

M03315-5G M03315-25G
FLUOM03315-5GEA 0 EUR
FLUOM03315-5G FLUOM03315-25G
Boc-DL-Pro-OH
Boc-DL-Pro-OH
Formel: C₁₀H₁₇NO₄
Molecular Weight: 215.25 g/mol
MDL: MFCD00520915
CAS-Nummer: 59433-50-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR