2-Azepanon, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
Azepan-2-one, 2-Oxohexamethyleneimine, 6-Aminocaproic acid lactam
M-Clarity™ Qualitätsstufe = MQ200
8.02800.0100
8.02800.0500
8.02800.1000
8.02800.9050
8.02800.0100EA
39.5
EUR
8.02800.0100
8.02800.0500
8.02800.1000
8.02800.9050
2-Azepanon, Sigma-Aldrich®
ε-Caprolactam
Formel:
C₆H₁₁NO Molecular Weight: 113,16 g/mol |
MDL:
MFCD00006936 CAS-Nummer: 105-60-2 EINECS: 203-313-2 |
Spezifikation Testergebnisse
Solubility | 4560 g/l (20 °C) |
Melting Point | 69.2 °C |
Molar Mass | 113.15 g/mol |
Bulk Density | 500 - 550 kg/m3 |
Boiling Point | 270.8 °C (1013 hPa) |
Vapor Pressure | 0.0014 hPa (20 °C) |
Flash Point | 152 °C DIN 51758 |
Explosion Limit | 1.4 - 8 %(V) |
Density | 1.01 g/cm3 (80 °C) |
pH | 7.0 - 8.5 (333 g/l, H2O, 20 °C) |
Ignition Point | 395 °C DIN 51794 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...