4-Formylmorpholin, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
Morpholine-4-carbaldehyde, N-Formylmorpholine
M-Clarity™ Qualitätsstufe = MQ200
8.18582.0100
8.18582.0500
8.18582.0100EA
36.4
EUR
8.18582.0100
8.18582.0500
4-Formylmorpholin, Sigma-Aldrich®
4-Formylmorpholin
Formel:
C₅H₉NO₂ Molecular Weight: 115.13 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006170 CAS-Nummer: 4394-85-8 EINECS: 224-518-3 |
Spezifikation Testergebnisse
Solubility | (20 °C) soluble |
Melting Point | 23 °C |
Molar Mass | 115.13 g/mol |
Boiling Point | 240 °C (1013 hPa) |
Vapor Pressure | 1 hPa (50 °C) |
Flash Point | 118 °C DIN 51758 |
Refractive Index | 1.484 (25 °C, 589 nm) |
Explosion Limit | 1.2 - 8.2 %(V) |
Density | 1.15 g/cm3 (25 °C) |
pH | 10 (250 g/l, H2O, 20 °C) |
Ignition Point | 345 °C DIN 51794 |
Synonyms | 4-Morpholinecarboxaldehyde |
Hill Formula | C5H9NO2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...