2,6-Dimethylbenzoesäure ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
A15014.03
A15014.06
A15014.14
A15014.03EA
40.2
EUR
A15014.03
A15014.06
A15014.14
2,6-Dimethylbenzoesäure ≥98%
2,6-Dimethylbenzoic acid
Purity: 98+%
CAS No.: 632-46-2
Molecular Formula: C9H10O2
Formula Weight: 150.18
Odour: Odourless
Melting Point: 114-117°
Beilstein Reference: 1863791
MDL No.: MFCD00002483
Formel:
C₉H₁₀O₂ Molecular Weight: 150.18 g/mol Siedepunkt: 155…160 °C (17 mmHg) Schmelzpunkt: 114…117 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002483 CAS-Nummer: 632-46-2 EINECS: 211-177-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...