Benzyldimethyldodecylammoniumbromid, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Benzyldodecyldimethylammonium bromide, Benzalkonium bromide, N-Benzyl-N,N-dimethyldodecan-1-aminium bromide
13373-10G-F
13373-50G-F
SIAL13373-10G-FEA
254
EUR
SIAL13373-10G-F
SIAL13373-50G-F
Benzyldimethyldodecylammoniumbromid, Sigma-Aldrich®
Benzyldimethyldodecylammoniumbromid
Formel:
C₂₁H₃₈BrN Molecular Weight: 384,44 g/mol |
MDL:
MFCD00011768 CAS-Nummer: 7281-04-1 EINECS: 230-698-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...