Benzyldimethyldodecylammoniumchlorid, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Benzyldimethyldodecylammonium chloride, N-Benzyl-N,N-dimethyldodecan-1-aminium chloride, Dodecyldimethylbenzylammonium chloride, Benzyldodecyldimethylammonium chloride dihydrate
13380-10G-F
13380-50G-F
SIAL13380-10G-FEA
208
EUR
SIAL13380-10G-F
SIAL13380-50G-F
Benzyldimethyldodecylammoniumchlorid, Sigma-Aldrich®
Benzyldimethyldodecylammoniumchlorid
Formel:
C₂₁H₃₈ClN Molecular Weight: 339,99 g/mol |
MDL:
MFCD00137276 CAS-Nummer: 139-07-1 EINECS: 205-351-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...