β-Chlor-4'-fluorpropiophenon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Chloro-1-(4-fluorophenyl)propan-1-one, 3-Chloro-4′-fluoropropiophenone, 3-Chloro-4'-fluoropropiophenone

135151-100G 135151-25G
SIAL135151-100GEA 148 EUR
SIAL135151-100G SIAL135151-25G
β-Chlor-4'-fluorpropiophenon, Sigma-Aldrich®
β-Chlor-4'-fluorpropiophenon
Formel: C₉H₈ClFO
Molecular Weight: 186,61 g/mol
MDL: MFCD00000991
CAS-Nummer: 347-93-3
EINECS: 206-475-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR