4-Chlor-7-nitrobenzofurazan, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
NBD chloride, 4-Chloro-7-Nitrobenz-2-Oxa-1,3-Diazole, NBD-Cl
M-Clarity™ Qualitätsstufe = MQ100
163260-5G
163260-1G
SIAL163260-5GEA
66.6
EUR
SIAL163260-5G
SIAL163260-1G
4-Chlor-7-nitrobenzofurazan, Sigma-Aldrich®
4-Chlor-7-nitrobenzofurazan
Formel:
C₆H₂ClN₃O₃ Molecular Weight: 199,55 g/mol |
MDL:
MFCD00005808 CAS-Nummer: 10199-89-0 EINECS: 233-496-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...