4-Methoxyphenethylamin, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
187305-100G
187305-25G
SIAL187305-100GEA
239
EUR
SIAL187305-100G
SIAL187305-25G
4-Methoxyphenethylamin, Sigma-Aldrich®
4-Methoxyphenethylamin
Formel:
C₉H₁₃NO Molecular Weight: 151,21 g/mol |
MDL:
MFCD00008192 CAS-Nummer: 55-81-2 EINECS: 200-245-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...