2-Brom-6-methoxynaphthalin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

200174-10G 200174-50G
SIAL200174-10GEA 38.5 EUR
SIAL200174-10G SIAL200174-50G
2-Brom-6-methoxynaphthalin, Sigma-Aldrich®
2-Bromo-6-methoxynaphthalene
Formel: C₁₁H₉BrO
Molecular Weight: 237,1 g/mol
MDL: MFCD00004062
CAS-Nummer: 5111-65-9
EINECS: 225-837-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR