tert-Butylnitrit, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ200
20370-100ML-F
20370-500ML-F
SIAL20370-100ML-FEA
86.8
EUR
SIAL20370-100ML-F
SIAL20370-500ML-F
tert-Butylnitrit, Sigma-Aldrich®
tert-Butylnitrit
Formel:
C₄H₉NO₂ Molecular Weight: 103.12 g/mol |
MDL:
MFCD00002055 CAS-Nummer: 540-80-7 EINECS: 208-757-0 UN: 2351 ADR: 3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...