2,6-Dimethylbenzoesäure
Lieferant: FLUOROCHEM
003427-5G
003427-100G
003427-25G
FLUO003427-5GEA
60.9
EUR
FLUO003427-5G
FLUO003427-100G
FLUO003427-25G
2,6-Dimethylbenzoesäure
2,6-Dimethylbenzoic acid
Formel:
C₉H₁₀O₂ Molecular Weight: 150,18 g/mol Siedepunkt: 155…160 °C (17 mmHg) Schmelzpunkt: 115…116 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002483 CAS-Nummer: 632-46-2 EINECS: 211-177-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...