tert-Butylnitrit, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

235385-100ML 235385-25ML
SIAL235385-100MLEA 87.8 EUR
SIAL235385-100ML SIAL235385-25ML
tert-Butylnitrit, Sigma-Aldrich®
tert-Butylnitrit
Formel: C₄H₉NO₂
Molecular Weight: 103.12 g/mol
MDL: MFCD00002055
CAS-Nummer: 540-80-7
EINECS: 208-757-0
UN: 2351
ADR: 3 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR