4-Chlor-7-nitrobenzofurazan, Sigma-Aldrich®

Lieferant: Merck

Synonyme: NBD chloride, 4-Chloro-7-Nitrobenz-2-Oxa-1,3-Diazole, NBD-Cl

M-Clarity™ Qualitätsstufe = MQ100

25455-25G 25455-5G
SIAL25455-25GEA 336 EUR
SIAL25455-25G SIAL25455-5G
4-Chlor-7-nitrobenzofurazan, Sigma-Aldrich®
4-Chlor-7-nitrobenzofurazan
Formel: C₆H₂ClN₃O₃
Molecular Weight: 199,55 g/mol
MDL: MFCD00005808
CAS-Nummer: 10199-89-0
EINECS: 233-496-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR