(1R,2R)-1,2-Diphenyl-1,2-ethandiol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
(1R,2R)-1,2-Diphenylethane-1,2-diol, (R,R)-(+)-1,2-Diphenyl-1,2-ethanediol
256277-25G
256277-5G
SIAL256277-25GEA
243
EUR
SIAL256277-25G
SIAL256277-5G
(1R,2R)-1,2-Diphenyl-1,2-ethandiol, Sigma-Aldrich®
(R,R)-(+)-Hydrobenzoin
Formel:
C₁₄H₁₄O₂ Molecular Weight: 214,26 g/mol |
MDL:
MFCD00064254 CAS-Nummer: 52340-78-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...