3-Methoxyphenethylamin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-(3-Methoxyphenyl)ethylamine

M-Clarity™ Qualitätsstufe = MQ100

270229-5G 270229-25G 270229-1G
SIAL270229-5GEA 53 EUR
SIAL270229-5G SIAL270229-25G SIAL270229-1G
3-Methoxyphenethylamin, Sigma-Aldrich®
3-Methoxyphenethylamin
Formel: C₉H₁₃NO
Molecular Weight: 151.21 g/mol
MDL: MFCD00008187
CAS-Nummer: 2039-67-0
EINECS: 218-017-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR