α,α-Dimethylpropiophenon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,2,2-Trimethylacetophenone

M-Clarity™ Qualitätsstufe = MQ100

280925-1G 280925-5G
SIAL280925-1GEA 75.3 EUR
SIAL280925-1G SIAL280925-5G
α,α-Dimethylpropiophenon, Sigma-Aldrich®
α,α-Dimethylpropiophenon
Formel: C₁₁H₁₄O
Molecular Weight: 162,23 g/mol
MDL: MFCD00008844
CAS-Nummer: 938-16-9
EINECS: 213-338-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR